missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gastrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599460
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Gastrin.
This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
Specifications
| Gastrin | |
| Polyclonal | |
| Unconjugated | |
| Gast | |
| Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14459, 2520, 25320 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P01350, P04563, P48757 | |
| Gast | |
| A synthesized peptide derived from human GAST(Accession P01350), corresponding to amino acid residues A70-N101. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction