missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516409
This item is not returnable.
View return policy
Description
PA5-16409 targets Gastrin in IHC (P) applications and shows reactivity with Human and Rat samples. The PA5-16409 immunogen is a Synthetic peptide from human gastrin.
This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
Specifications
| Gastrin | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P01350, P04563 | |
| Gast | |
| A Synthetic peptide from human gastrin | |
| 500 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Gast | |
| Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin | |
| Rabbit | |
| purified | |
| RUO | |
| 2520, 25320 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction