missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Mouse anti-Human, Clone: GAST/2635, Novus Biologicals™
Mouse Monoclonal Antibody
£205.00
Specifications
| Antigen | Gastrin |
|---|---|
| Clone | GAST/2635 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 1-5 ug/ml |
| Applications | ELISA |
Description
Gastrin Monoclonal specifically detects Gastrin in Human samples. It is validated for ELISA.Specifications
| Gastrin | |
| 0.2 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| GAS, gastrin | |
| Recombinant full-length human Gastrin protein | |
| Primary | |
| Store at 4°C. |
| GAST/2635 | |
| ELISA 1-5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2520 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title