missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Mouse anti-Human, Clone: GAST/2634, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07821-100ug
This item is not returnable.
View return policy
Description
Gastrin Monoclonal specifically detects Gastrin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications
| Gastrin | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 1-5 μg/mL, Immunohistochemistry-Paraffin | |
| GAS, gastrin | |
| Recombinant full-length human Gastrin protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| GAST/2634 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2520 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction