missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gastrin Monoclonal Antibody (C25), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA551080

 View more versions of this product

Product Code. 17910123

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gastrin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P01350, P48757
Gast
Recombinant Human GT protein, Met1-Asn101 (Accession #P01350).
20 μL
Primary
Human, Mouse
Antibody
IgG2a κ
Western Blot
C25
Unconjugated
Gast
Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin
Mouse
Protein A/G
RUO
14459, 2520
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.