missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Antibody (GAST/2631), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08657AF594
This item is not returnable.
View return policy
Description
Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for ELISA
Specifications
| Gastrin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| GAS, gastrin | |
| Recombinant full-length human Gastrin protein (Uniprot: P01350) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Cell Biology, Immunology, Signal Transduction | |
| 2520 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA | |
| GAST/2631 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction