missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Antibody (c3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43361
This item is not returnable.
View return policy
Description
Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Serum,Immunocytochemistry/ Immunofluorescence
Specifications
| Gastrin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.2-2 μg/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation, Serum | |
| GAS, gastrin | |
| Immunized with OVA - Gastrin (Uniprot#: P01350) | |
| 100 μL | |
| Cancer, Cardiovascular Biology, Cell Biology, Immunology, Signal Transduction | |
| 2520 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| c3 | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction