missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastric Lipase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97330-100ul
This item is not returnable.
View return policy
Description
Gastric Lipase Polyclonal antibody specifically detects Gastric Lipase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Gastric Lipase | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:10000 | |
| EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
| Produced in rabbits immunized with E. coli-derived Human Gastric Lipase fragment. | |
| 100 μg | |
| Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
| 8513 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction