missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GASP2 Monoclonal Antibody (289802)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523992
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
The GASP2 gene encodes for a protein known as G protein-coupled receptor-associated sorting protein 2. GASP2 is involved in the regulation of G protein-coupled receptors (GPCRs), which are crucial for transmitting signals from the external environment to the inside of a cell. This protein plays a key role in the intracellular trafficking and sorting of GPCRs, affecting their recycling and degradation pathways, which are critical for receptor resensitization and downregulation. GASP2 assists in marking GPCRs for degradation post endocytosis, thereby influencing receptor availability on the cell surface and modulating cellular responsiveness to ligands. Dysregulation of GASP2 expression or function can impact GPCR signaling pathways, potentially affecting various physiological processes and contributing to disease states where GPCR signaling is altered, such as in certain neurological disorders and cancers. The role of GASP2 in GPCR regulation makes it a point of interest for therapeutic targeting to modulate GPCR-related signaling pathways for clinical benefits in diverse diseases.
Specifications
| GASP2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96NZ8 | |
| WFIKKN1 | |
| Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 289802 | |
| Unconjugated | |
| WFIKKN1 | |
| C16orf12; GASP2; GASP-2; Growth and differentiation factor-associated serum protein 2; hGASP-2; PSEC0040; RJD2; WAP four-disulfide core domain 20A; WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing protein; WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1; WAP, FS, Ig, KU, and NTR-containing protein; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 1; WFDC20A; WFIKKN; WFIKKN1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 117166 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction