missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gasdermin-C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00
Specifications
| Antigen | Gasdermin-C |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Gasdermin-C Polyclonal specifically detects Gasdermin-C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Gasdermin-C | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56169 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LPVGRIEEPFWQNFKHLQEEVFQKIKTLAQLSKDVQDVMFYSILAMLRDRGALQDLMNMLELDSSGHLDGPGGAILKKLQQDSNHAWFNPKDP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| gasdermin C, gasdermin-C, Melanoma-derived leucine zipper-containing extranuclear factor, MLZEmelanoma-derived leucine zipper, extra-nuclear factor | |
| GSDMC | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title