missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103864
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GAS8.
This gene includes 11 exons spanning 25 kb and maps to a region of chromosome 16 that is sometimes deleted in breast and prostrate cancer. The second intron contains an apparently intronless gene, C16orf3, that is transcribed in the opposite orientation. This gene is a putative tumor suppressor gene.
Specifications
| GAS8 | |
| Polyclonal | |
| Unconjugated | |
| GAS8 | |
| beta growth arrest specific protein 11; CILD33; DRC4; Dynein regulatory complex subunit 4; Gas11; GAS-11; Gas8; GAS-8; growth arrest specific 11; growth arrest specific 8; growth arrest specific protein 11; growth arrest-specific protein 11; Growth arrest-specific protein 8; MGC138326 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 104346, 2622, 361438 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95995, Q499U4, Q60779 | |
| GAS8 | |
| A synthesized peptide derived from human GAS8(Accession O95995), corresponding to amino acid residues L165-K215. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction