missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS8 Antibody (OTI10G2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70795B
This item is not returnable.
View return policy
Description
GAS8 Monoclonal specifically detects GAS8 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GAS8 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GAS8 | |
| Full length recombinant protein of human GAS8 (NP_001472) produced in E.coli. | |
| 0.1 mL | |
| 2622.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI10G2 | |
| Western Blot | |
| GAS-11, GAS11MGC138326, GAS-8, growth arrest specific 11, growth arrest-specific 11, growth arrest-specific 8, Growth arrest-specific protein 11, growth arrest-specific protein 8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction