missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS7 Antibody (OTI2F6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71376CL1
This item is not returnable.
View return policy
Description
GAS7 Monoclonal antibody specifically detects GAS7 in Canine,Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| GAS7 | |
| Monoclonal | |
| CoraFluor 1 | |
| GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
| Full length human recombinant protein of human GAS7 (NP_003635) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8522 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI2F6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction