missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579300
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. IHC: mouse spleen tissue, rat spleen tissue. ICC/IF: A431 cell. Flow: THP-1 cell.
This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| GAS6 | |
| Polyclonal | |
| Unconjugated | |
| GAS6 | |
| AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14456, 2621, 58935 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q14393, Q61592, Q63772 | |
| GAS6 | |
| E.coli-derived human GAS 6 recombinant protein (Position: M488-S660). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction