Learn More
Invitrogen™ GAS6 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMGAS6X5
Additional Details : Weight : 0.33800kg
Description
The Mouse GAS6 ELISA quantitates Ms GAS6 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms GAS6. Principle of the method The Mouse GAS6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Growth arrest specific proteins, including Gas1 and Gas6, are activated in quiescent cells. Gas1-induced growth arrest is mediated by p53, and Gas1 appears to be able to suppress tumor cell growth. Gas6, a ligand for the tyrosine kinase receptor Axl, was initially identified as a member of the vitamin K-dependent protein family and exhibits a high degree of amino acid sequence homology to protein S, a negative co-regulator in the coagulation pathway.Specifications
Q61592 | |
20 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
14456 | |
<12% | |
SAV-HRP, Adhesive Plate Covers, Standard, Pre-coated 96 well plates, Assay Diluent Concentrate, Chromogen, Biotinylated Detection Antibody, Wash Buffer, Stop Solution | |
5 x 96 Tests | |
Immunology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
20.48 pg/mL to 5000 pg/mL | |
20 pg/mL to 5000 pg/mL | |
Biotin | |
Kinase & Inhibitors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
Gas-6 | |
<10% | |
HRP | |
RUO | |
Plasma, 10 μ;L; Serum, 10 μ;L; Supernatant, 100 μ;L | |
Gas-6 | |
1 hr. 20 min. |