missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAS6 Monoclonal Antibody (100140)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561975

Product Code. 30533684

  • £340.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GAS6
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q14393
GAS6
Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678.
100 μg
Primary
Human
Antibody
IgG1
ELISA
100140
Unconjugated
GAS6
AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor
Mouse
Protein A/G
RUO
2621
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.