missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gas6 Antibody (RM0084-6J2), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21724AF488
This item is not returnable.
View return policy
Description
Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Gas6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GAS6 | |
| Mouse recombinant Gas6 | |
| 0.1 mL | |
| 2621 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, ELISA | |
| RM0084-6J2 | |
| Western Blot, ELISA | |
| AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction