missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gas6 Antibody (100127) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB885A
This item is not returnable.
View return policy
Description
Gas6 Monoclonal antibody specifically detects Gas6 in Human samples. It is validated for Western Blot
Specifications
| Gas6 | |
| Monoclonal | |
| APC | |
| AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession # NP_000811 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 100127 | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2621 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction