missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528935
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (91%), Xenopus laevis (95%), Chicken (97%), Sheep (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GAS41 has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors.
Specifications
| GAS41 | |
| Polyclonal | |
| Unconjugated | |
| YEATS4 | |
| 4930573H17Rik; B230215M10Rik; GAS41; Glioma-amplified sequence 41; glioma-amplified sequence-41; nuBI1; NuBI-1; NuMA binding protein 1; nuMA-binding protein 1; RGD1305741; YAF9; YEATS domain containing 4; YEATS domain-containing protein 4; Yeats4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95619 | |
| YEATS4 | |
| Full length human GAS41 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction