missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS41 Polyclonal Antibody

GAS41 Polyclonal Antibody
Brand: Invitrogen PA599230
This item is not returnable.
View return policy
Description
GAS41 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| GAS41 | |
| Polyclonal | |
| Unconjugated | |
| YEATS4 | |
| 4930573H17Rik; B230215M10Rik; GAS41; Glioma-amplified sequence 41; glioma-amplified sequence-41; nuBI1; NuBI-1; NuMA binding protein 1; nuMA-binding protein 1; RGD1305741; YAF9; YEATS domain containing 4; YEATS domain-containing protein 4; Yeats4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64050, 8089 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95619, Q9CR11 | |
| YEATS4 | |
| A synthesized peptide derived from human YEATS4(Accession O95619), corresponding to amino acid residues V63-T113. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction