missing translation for 'onlineSavingsMsg'
Learn More

GAS41 Antibody, Novus Biologicals™

Product Code. 18327038 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327038 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327038 Supplier Novus Biologicals Supplier No. H00008089B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GAS41 Polyclonal antibody specifically detects GAS41 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS41
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_006521.1
Gene Alias B230215M10Rik, Gas41,4930573H17Rik, GAS41glioma-amplified sequence-41, Glioma-amplified sequence 41, NuBI1, NuBI-1, NuMA binding protein 1, NuMA-binding protein 1, YAF9, YEATS domain containing 4, YEATS domain-containing protein 4
Host Species Mouse
Immunogen YEATS4 (NP_006521.1, 1 a.a. - 227 a.a.) full-length human protein. MFKRMAEFGPDSGGRVKGVTIVKPIVYGNVARYFGKKREEDGHTHQWTVYVKPYRNEDMSAYVKKIQFKLHESYGNPLRVVTKPPYEITETGWGEFEIIIKIFFIDPNERPVTLYHLLKLFQSDTNAMLGKKTVVSEFYDEMIFQDPTAMMQQLLTTSRQLTLGAYKHETEFAELEVKTREKLEAAKKKTSFEIAELKERLKASRETINCLKNEIRKLEEDDQAKDI
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8089
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.