missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS2L3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30359PE
This item is not returnable.
View return policy
Description
GAS2L3 Polyclonal antibody specifically detects GAS2L3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| GAS2L3 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| GAS2-like protein 3, growth arrest-specific 2 like 3, Growth arrest-specific protein 2-like 3 | |
| Maps to a region between residue 325 to 375 of human Growth Arrest-Specific 2 Like 3 using the numbering given in entry NP_777602.1 (GeneID 283431). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 283431 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction