missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GAS2L3 Polyclonal antibody specifically detects GAS2L3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | GAS2L3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GAS2-like protein 3, growth arrest-specific 2 like 3, Growth arrest-specific protein 2-like 3 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 644 to 694 of human Growth Arrest-Specific 2 Like 3 using the numbering given in entry NP_777602.1 (GeneID 283431). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?