missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS2 Monoclonal Antibody (K94023_17D9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556356
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sequence of this protein is as follows: SGPGLSDMHQ YSQWLASRHE ANLLPMKEDL ALWLTNLLGK EITAETFMEK LDNGALLCQL AETMQEKFKE SMDANKPTKN LPLKKIPCKT SAPSGSFFAR DNTANFLSWC RDLGVDETCL FESEGLVLHK QPREVCLCLL ELGRIAARYG VEPPGLIKLE KEIEQEETLS
Gas2 is a 313 amino acid protein encoded by the human gene GAS2. Gas2 is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 and caspase-7) at Asparagine 278 during apoptosis. The cleaved form resulting from this dramatically induces the rearrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is believed to also be involved in the membrane ruffling process. During the G0-G1 transition phase Gas2 can be found phosphorylated on its serine residues. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated but when hyperphosphorylated it will accumulate at membrane ruffles. Gas2 is specifically expressed at growth arrest and is ubiquitously expressed with highest levels found in liver, lung and kidney.
Specifica
| GAS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O43903 | |
| GAS2 | |
| Recombinant human GAS2 (immunogen range is 11-180). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K94023_17D9 | |
| Unconjugated | |
| GAS2 | |
| GAS 2; Gas2; Gas-2; growth arrest specific 2; growth arrest-specific 2; growth arrest-specific protein 2; RGD1563167 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto