missing translation for 'onlineSavingsMsg'
Learn More

Gas1 Antibody (11C47), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18410168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410168 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410168 Supplier Novus Biologicals Supplier No. NBP222032MFV450

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rat Monoclonal Antibody

Gas1 Monoclonal antibody specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Gas1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 11C47
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
Host Species Rat
Immunogen Human recombinant Gas1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2619
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.