Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head without Filter Disc
Gas washing head for bottles acc. to DIN 12463
Brand:  Lenz Laborglasintrumente™ 05500610
Additional Details : Weight : 0.10000kg
Product Code. 10676830
  • £40.00 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596
  • With cone NS 29/32
  • Material: DURAN™ tubing
Specifications
Specifications
With thread connections (GL 14): 11 mm
Gas washing bottles acc. to DIN 12463
1