Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head with Filter Disc
Gas washing head for bottles acc. to DIN 12463
£58.00 - £65.00
Specifications
Material | Duran Tubing |
---|---|
For Use With (Equipment) | Gas washing bottles acc. to DIN 12463 |
Quantity | 1 |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10508431
|
Lenz Laborglasintrumente™
05510402 |
Olive Connections |
£58.00
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
11973551
|
Lenz Laborglasintrumente™
05510411 |
GL 14 connections |
£65.00
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12596
- With cone NS 29/32
- Material: DURAN™ tubing
Specifications
Duran Tubing | |
Gas washing bottles acc. to DIN 12463 | |
1 |