missing translation for 'onlineSavingsMsg'
Learn More

3M™ Gas, Vapor, and Particulate Filter, 6000 Series

Product Code. 11872863
Change view
Click to view available options
Model: 6099; Filter type: ABEK2P3R
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11872863 Model: 6099; Filter type: ABEK2P3R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11872863 Supplier 3M™ Supplier No. 7100104327

Please to purchase this item. Need a web account? Register with us today!

Edge

Provides excellent field of vision as a result of the unique trapezoidal shape. The 3M Gas, Vapor, and Particulate Filter uses bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks.

Unique, compatible Bayonet Filter Attachment System allows different types of filters to just be clicked into place ensuring optimum ease of use.

  • Lightweight, low breathing resistance, and well-balanced when fitted to mask
  • Provides excellent field of vision as a result of the unique trapezoidal shape
  • Bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Organic Vapor
Certifications/Compliance EN14387:2004 + A1:2008,‎ CE Approved
Color Gray, Brown, White, Yellow, Green
Filter Type A2B2E2K2HgP3, A2B2E2K2HgP3R
Product Type Gas, Vapor and Particulate Filter
For Use With (Application) Typical industrial applications for filters are laboratories and particulate applications
For Use With (Equipment) 3M™ 6000 and 3M™ 7907 Series Full Face Masks
Model 6099
Quantity 4/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.