Learn More
3M™ Gas, Vapour and Particulate Filter
Description
Unique, compatible Bayonet Filter Attachment System allows different types of filters to just be clicked into place ensuring optimum ease of use.
- Lightweight, low breathing resistance, and well-balanced when fitted to mask
- Provides excellent field of vision as a result of the unique trapezoidal shape
- Bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks
Specifications
Specifications
| Cartridge Type | P3 R |
| Filter Type | FFP3 |
| Product Type | Gas, Vapor and Particulate Filter |
| For Use With (Application) | Typical industrial applications for filters are laboratories and particulate applications |
| For Use With (Equipment) | 3M™ 6000 and 3M™ 7907 Series Full Face Masks |
| Model | 6035 |