Learn More
3M™ Gas & Vapor Filter GF32 AX
Product Code. 17756983
Change view
Click to view available options
Filter; Model: DT-4007E
Product Code.
17756983 Filter; Model: DT-4007E
1 options
Product Code. 17756983 Supplier 3M™ Supplier No. 7100264836
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Gas & Vapor Filter DT-4007E is a DIN 40 threaded, replaceable filter offering AX protection against certain low-boiling point organic vapors.
  • AX protection against certain low-boiling point organic Vapors
  • DIN40 threaded connection
  • Compatible with 3M™ Reusable Half Mask HF-300 Series, 3M™ Reusable Full Face Masks FF-300 Series and FF-600 Series
  • Compatible with 3M™ Powered Air Respirator PF-600E Series
TRUSTED_SUSTAINABILITY
Specifications
Product Type Filter
For Use With (Equipment) 3M™ Reusable Half Mask HF-300 Series, Reusable Full Face Masks FF-300 Series and FF-600 Series, Powered Air Respirator PF-600E Series
Model DT-4007E
Quantity 10/Cs.