Learn More
3M™ Gas and Vapour Filter GF22 A2B2, DT-4005E, 10 Each/Case
Description
- Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
- Features a DIN40 mm threaded connection
- Please note DT-Series filters were previously known as Pro2000 Filters
- To identify which filter is suitable for your environment, and to aid in the implementation of a filter changeout schedule, please use the 3M Service Life and Select Software, available from www.3m.com/sls.
Specifications
Specifications
| Product Type | Filter |
| For Use With (Equipment) | 3M™ Powered Air Respirator PF-600E Series |
| Model | DT-4005E |
| Quantity | 10/Cs. |