Learn More
BINDER™ Gas-tight Option Model
Product Code. 16454039 Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 16454039
Brand: BINDER™ 80121044
Only null left
Add to basket
Add to basket
For Use With: BF 56
  • Includes additional measures for the greatest possible impermeability to gas
  • Not in conjunction with access port or interior lighting option
Specifications
Description Gas-tight option model - Incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option
For Use With (Equipment) BINDER Chambers BF 56
Material Steel
Type Gas-tight Option Model