Learn More
BINDER™ Gas-tight Option Model
Product Code. 17807108 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
M 115 E3.1 Drying/Heating Oven
M 260 E3.1 Drying/Heating Oven
M 56 E3.1 Drying/Heating Oven
Unit Size:
Each
Product Code. 17807108
Brand: BINDER™ 80122308
Only null left
Add to basket
Add to basket
Gas-tight Option Model including additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option.
  • Accessory
Specifications
For Use With (Equipment) M 56 E3.1 Drying/Heating Oven
Material Aluminum
Type Gas-tight Option Model