missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Gas Distribution Tube, Type A

Product Code. 16892986
Change view
Click to view available options
Pore Size:
0
1
2
3
4
Diameter (Metric) Inner Tubing:
25 mm
34 mm
Unit Size:
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Inner Tubing unitSize
16892986 4 34 mm Pack of 5
16812986 1 25 mm Pack of 5
16802986 0 25 mm Pack of 5
16852986 0 34 mm Pack of 5
16842986 4 25 mm Pack of 5
16872986 2 34 mm Pack of 5
16882986 3 34 mm Pack of 5
16832986 3 25 mm Pack of 5
16862986 1 34 mm Pack of 5
16822986 2 25 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 16892986 Supplier ROBU VitraPOR™ Supplier No. 19344
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gas Distribution Tube with centered bowl.

  • For fine dispersion of gases in liquids
  • Highly resistant to most chemicals and temperatures up to 500°C

Specifications

Material Borosilicate Glass (3.3)
Type Type A
Pore Size 4
Diameter (Metric) Inner Tubing 34 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.