missing translation for 'onlineSavingsMsg'
Learn More

Gas Detector Tube

Product Code. 10179640
Change view
Click to view available options
Measurement Range:
5 to 23
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10179640 5 to 23 Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10179640 Supplier Draeger Safety Supplier No. 6728081
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Draeger tubes are the most widely used form of spot measurement in industry

Gas detector pump that is independent of external energy sources and therefore there are no usage restrictions in areas of explosive hazards.
Dräger tubes are the most widely used form of spot measurement in industry. More than 500 substances can be detected and measured using the system. Typical applications include: COSHH monitoring, emission measurements, quality control, process control, compressed breathing air testing, as well as environmental and emergency response monitoring. Gas is drawn into the tube using the Accuro bellows pump where it reacts with chemical on a carrier material. In the presence of the target gas a colour change occurs and the length of that discolouration is proportional to the concentration. The concentration can be determined from the calibrated printed scale on the outside of the tube.

Specifications

Components Measured Oxygen
For Use With (Equipment) Dräger Aerotest
Measurement Range 5 to 23
Quantity 8 Pk.
Response Time 1min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.