missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GART Recombinant Rabbit Monoclonal Antibody (23GB5520), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552873
This item is not returnable.
View return policy
Description
MERTK is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including LGALS3, TUB, TULP1 or GAS6. Regulates many physiological processes including cell survival, migration, differentiation, and phagocytosis of apoptotic cells (efferocytosis). Ligand binding at the cell surface induces autophosphorylation of MERTK on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by ligand, interacts with GRB2 or PLCG2 and induces phosphorylation of MAPK1, MAPK2, FAK/PTK2 or RAC1. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium (RPE) as a regulator of rod outer segments fragments phagocytosis. Plays also an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.
Specifications
| GART | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GART | |
| 5'-phosphoribosylglycinamide transformylase; AIR synthase; AIRS; Gaps; GAR transformylase; GARS; Gart; GARTF; glycinamide ribonucleotide formyltransferase; Glycinamide ribonucleotide synthetase; MGC47764; PAIS; PGFT; Phosphoribosylamine--glycine ligase; Phosphoribosyl-aminoimidazole synthetase; Phosphoribosylformylglycinamidine cyclo-ligase; Phosphoribosylglycinamide formyltransferase; phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase; Phosphoribosylglycinamide synthetase; PRGS; Trifunctional purine biosynthetic protein adenosine-3; Trifunctional purine biosynthetic protein adenosine-3-like protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14450, 2618, 288259 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5520 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22102, Q64737 | |
| GART | |
| A synthesized peptide derived from human GART (550-750AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction