Learn More
GART Mouse anti-Human, Unconjugated, Clone: 2A11E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67939-1-IG-150UL
This item is not returnable.
View return policy
Description
The GART antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GART. This antibody recognizes human, mouse, rat antigen. The GART antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GART | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 2618 | |
| IgG1 |
| Western Blot, ELISA | |
| 2A11E2 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |