missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GART Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32207CL1
This item is not returnable.
View return policy
Description
GART Polyclonal antibody specifically detects GART in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| GART | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| AIRS, EC 6.3.3.1, EC 6.3.4.13, GARS, GARTF, MGC47764, PGFTEC 2.1.2.210PAIS, phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamidesynthetase, phosphoribosylaminoimidazole synthetase, trifunctional purine biosynthetic protein adenosine-3 | |
| The immunogen this antibody was made to, maps to a region between residue 960 to 1010 of human Glycinamide Ribonucleotide Synthetase using the numbering given in entry NP_000810.1 (GeneID 2618). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2618 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction