missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GARS Monoclonal Antibody (1641CT125.1.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537709
This item is not returnable.
View return policy
Description
GARS Monoclonal Antibody for Western Blot, Flow
GARS is a glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. This protein is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. The protein has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.
Specifications
| GARS | |
| Monoclonal | |
| Unconjugated | |
| GARS1 | |
| 6.1.1.14; AIR synthase; AIRS; Ap4A synthetase; AP-4-A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; CMT2D; Diadenosine tetraphosphate synthetase; DSMAV; EC 2.1.2.29; GAR transformylase; GARS; gars1; GART; GARTF; Gena201; GENA202; glycine tRNA ligase; glycine--tRNA ligase; glycyl-tran synthetase; glycyl-tRNA synthetase; glycyl-tRNA synthetase 1; glycyl-tRNA synthetase-like protein; glyRS; HMN5; metronome; MGC47764; mrn; Nmf249; PAIS; PGFT; PRGS; RGD1559871; Sgrp23; si:dkey-276i5.1; SMAD1; storage granule protein 23; Unknown (protein for IMAGE:7254992); wu:fb02f03 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1641CT125.1.1 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P41250 | |
| GARS, GARS1 | |
| recombinant protein between 35-305 amino acids from human GARS. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction