missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARS Antibody (4E10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42679C
Additional Details : Weight : 0.00970kg
Description
GARS Monoclonal specifically detects GARS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GARS | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GARS | |
Recombinant human GARS (43-289aa) purified from E. coli | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
2617 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
40000000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |