missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GARP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584380
This item is not returnable.
View return policy
Description
Immunogen sequence: GNPLSCCGNG WLAAQLHQGR VDVDATQDLI CRFSSQEEVS LSHVRPEDCE K Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 96%.
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.
Specifications
| GARP | |
| Polyclonal | |
| Unconjugated | |
| LRRC32 | |
| AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14392 | |
| LRRC32 | |
| Recombinant protein corresponding to Human LRRC32. Recombinant protein control fragment (Product #RP-103601). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction