missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARP Monoclonal Antibody (YGIC86), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 67-9891-80
This item is not returnable.
View return policy
Description
Rat Monoclonal Antibody
GARP Monoclonal antibody specifically detects GARP in Mouse samples. It is validated for Flow Cytometry
Specifications
| GARP | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; GARP; Garpin; Glycoprotein A repetitions predominant; Leucine-rich repeat-containing protein 32; LRRC32 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| YGIC86 | |
| Super Bright 702 | |
| LRRC32 | |
| LRRC32 | |
| Affinity chromatography | |
| RUO | |
| 434215 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction