missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GARP Monoclonal Antibody (YGIC86), Super Bright™ 702, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
Brand: Invitrogen™ 67-9891-82
This item is not returnable.
View return policy
Description
Rat Monoclonal Antibody
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.
Specifications
| GARP | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| G3XA59 | |
| LRRC32 | |
| Affinity chromatography | |
| RUO | |
| 434215 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| YGIC86 | |
| Super Bright 702 | |
| LRRC32 | |
| AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction