missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARP Monoclonal Antibody (YGIC86), Super Bright™ 436, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£132.00 - £370.00
Specifications
| Antigen | GARP |
|---|---|
| Clone | YGIC86 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predomit (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray anal
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.Specifications
| GARP | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 434215 | |
| Primary | |
| Antibody |
| YGIC86 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Rat | |
| Mouse | |
| IgG2a κ | |
| 4° C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title