missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gardiquimod diTFA, MedChemExpress
Description
Gardiquimod diTFA, an imidazoquinoline analog, is a TLR7/8 agonist. Gardiquimod diTFA could inhibit HIV-1 infection of macrophages and activated peripheral blood mononuclear cells (PBMCs). Gardiquimod diTFA specifically activates TLR7 when used at concentrations below 10 μM.
Specifications
Specifications
| Chemical Name or Material | Gardiquimod diTFA |
| CAS | 1159840-61-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H25F6N5O5 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 50 mg/mL (92.35 mM; Need ultrasonic) ∣H2O : 25 mg/mL (46.17 mM; Need ultrasonic) |
| SMILES | OC(C)(C)CN1C(CNCC)=NC2=C1C3=CC=CC=C3N=C2N.FC(C(O)=O)(F)F.FC(C(O)=O)(F)F |
| Molecular Weight (g/mol) | 541.44 |
| Formula Weight | 541.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?