missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAPDHS Monoclonal Antibody (Hs-8), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544094
This item is not returnable.
View return policy
Description
This antibody reacts with GAPDHS, the sperm-specific glyceraldehyde phosphate dehydrogenase, which is an intra-acrosomal protein.
Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. Because GAPDH is a protein expressed in large amounts and which is required at all times for important 'house keeping' functions, levels of GAPDH mRNA are often measured and used as standards in studies of mRNA expression. Increasingly, scientists are making use of specific antibodies to GAPDH in comparable studies of levels of protein expression. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer's disease, and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington's disease. Associations with actin and tubulin have also been reported. The protein may also have a role in the regulation of apoptosis, and interestingly migrates from the cytoplasm into the nucleus when cells become apoptotic.
Specifications
| GAPDHS | |
| Monoclonal | |
| 0.06 mg/mL | |
| TBS with 0.2% BSA and 15mM sodium azide; pH 8 | |
| O14556 | |
| GAPDHS | |
| Freshly ejaculated human sperms. | |
| 50 Tests | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgM |
| Flow Cytometry | |
| Hs-8 | |
| FITC | |
| GAPDHS | |
| epididymis secretory protein Li 278; GAPD2; GAPDH S; GAPDH2; gapdh-2; GAPDHS; GAPDS; Gapd-s; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; HEL-S-278; HSD35; HSD-35; LOW QUALITY PROTEIN: glyceraldehyde-3-phosphate dehydrogenase, testis-specific; spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2; Spermatogenic glyceraldehyde-3-phosphate dehydrogenase | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 100517097, 26330 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction