missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH (Human Specific) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80570-1-RR-100UL
This item is not returnable.
View return policy
Description
The GAPDH (Human Specific) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human GAPDH (Human Specific). This antibody recognizes human antigen. The GAPDH (Human Specific) antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| GAPDH (Human Specific) | |
| Recombinant Monoclonal | |
| 400 μg/mL | |
| G3PD; GAPD; MGC88685 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2G21 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 2597 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction