missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GAPDH Monoclonal antibody specifically detects GAPDH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | GAPDH |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | BLR103H |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide |
| Gene Alias | aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH |
| Host Species | Rabbit |
| Immunogen | between 225 and 275 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?