missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Antibody (1A10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications

£184.00 - £400.00

Specifications

Antigen GAPDH
Clone 1A10
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 5 ug/ml, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18064105
View Documents
Novus Biologicals
NBP1-47339SS
0.025 mL
£184.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18759953
View Documents
Novus Biologicals
NBP1-47339
£400.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GAPDH
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
P04406
2597
GAPDH antibody developed against purified recombinant fragment of human GAPDH expressed in E. coli. [UniProt# P04406]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1A10
Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 5 ug/ml, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Monoclonal
Precipitation
RUO
Human, Mouse
aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
GAPDH
IgG1
Ammonium sulfate precipitation
36 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.