missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Antibody (13H12) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27103JF669
This item is not returnable.
View return policy
Description
GAPDH Monoclonal antibody specifically detects GAPDH in Drosophila,Human,Monkey,Mouse,Primate,Rat,Sheep samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/ Immunofluorescence
Specifications
| GAPDH | |
| Monoclonal | |
| Janelia Fluor 669 | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| Amino acids between 275 and 325 of glyceraldehyde 3-phosphate dehydrogenase protein were used as the immunogen for this GAPDH antibody. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, Chromatin Research, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| 2597 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, Immunocytochemistry/Immunofluorescence | |
| 13H12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Drosophila, Human, Monkey, Mouse, Primate, Rat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction